1-(2,4-Dimethylphenyl)-2-thiourea 98%

Danger

Synonyms: 2,4-DimethylphenylthioureaN-(2,4-Dimethylphenyl)thiourea1-(2,4-Dimethylphenyl)thiourea1-(2,4-Xylyl)thiourea

L12206-06 L12206-03 L12206-03 L12206-06
AAAL12206-03EA 175.46 USD
AAAL12206-03 CAAAAL12206-06 AAAL12206-06 CAAAAL12206-03
1-(2,4-Dimethylphenyl)-2-thiourea 98%
1-(2,4-Dimethylphenyl)-2-thiourea
Formula: (C₆H₃(CH₃)₂)NC(S)NH₂
MW: 180.27 g/mol
Melting Pt: 177…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00041164
CAS Number: 16738-20-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR