Dipropyl sulfite 99%

Warning

L12033-06 L12033-14 L12033-14 L12033-06
AAAL12033-06EA 26.38 USD
CAAAAL12033-14 AAAL12033-06 CAAAAL12033-06 AAAL12033-14
Dipropyl sulfite 99%
Dipropyl sulphite
Formula: C₆H₁₄O₃S
MW: 166.24 g/mol
Boiling Pt: 107…109 °C/42 mm
Density: 1.030
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059404
CAS Number: 623-98-3
UN: 3334
ADR: "9",

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR