1-(2-Methoxyphenyl)-2-thiourea 94%

Danger

Synonyms: 2-MethoxyphenylthioureaN-(2-Methoxyphenyl)thiourea1-(2-Methoxyphenyl)thiourea

L11840-06 L11840-06
AAAL11840-06EA 46.1 USD
CAAAAL11840-06 AAAL11840-06
1-(2-Methoxyphenyl)-2-thiourea 94%
1-(2-Methoxyphenyl)-2-thiourea
Formula: (C₆H₄(OCH₃))NC(S)NH₂
MW: 182.25 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00022162
CAS Number: 1516-37-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR