2-Methoxy-5-nitrophenol 98%

Warning

Synonyms: 2-Hydroxy-4-nitroanisole5-Nitroguaiacol

L11822-18 L11822-09 L11822-09 L11822-18 L11822-30 L11822-30
AAAL11822-18EA 163.89 USD
CAAAAL11822-18 AAAL11822-18 CAAAAL11822-09 AAAL11822-30 AAAL11822-09 CAAAAL11822-30
2-Methoxy-5-nitrophenol 98%
2-Methoxy-5-nitrophenol
Formula: C₇H₇NO₄
MW: 169.14 g/mol
Boiling Pt: 110…112 °C/1 mm
Melting Pt: 103…106 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015561
CAS Number: 636-93-1
EINECS: 211-269-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR