2-(Trimethylsilyl)benzothiazole tech. 90%, Technical Grade

Warning

L11625-06 L11625-03 L11625-06 L11625-03
AAAL11625-06EA 208.68 USD
AAAL11625-06 CAAAAL11625-03 CAAAAL11625-06 AAAL11625-03
2-(Trimethylsilyl)benzothiazole tech. 90%, Technical Grade
2-(Trimethylsilyl)benzothiazole
Formula: C₆H₁₁NSSi
Boiling Pt: 128…130 °C/2 mm
Density: 1.06
Storage Temperature: Ambient
MDL Number: MFCD00068185
CAS Number: 32137-73-8
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR