3-Nitrophenyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L11335-14 L11335-03 L11335-03 L11335-14
AAAL11335-03EA 35.09 USD
CAAAAL11335-14 CAAAAL11335-03 AAAL11335-03 AAAL11335-14
3-Nitrophenyl isothiocyanate 97%
3-Nitrophenyl isothiocyanate
Formula: C₇H₄N₂O₂S
MW: 180.19 g/mol
Melting Pt: 56…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00041245
CAS Number: 3529-82-6
EINECS: 000-000-0
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR