1-(2,4-Dichlorophenyl)-2-thiourea 98+%

Danger

Synonyms: 2,4-DichlorophenylthioureaN-(2,4-Dichlorophenyl)thiourea1-(2,4-Dichlorophenyl)thiourea

L11314-06 L11314-06
AAAL11314-06EA 79.23 USD
CAAAAL11314-06 AAAL11314-06
1-(2,4-Dichlorophenyl)-2-thiourea 98+%
1-(2,4-Dichlorophenyl)-2-thiourea
Formula: (C₆H₃Cl₂)NC(S)NH₂
MW: 221.11 g/mol
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00022161
CAS Number: 6326-14-3
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR