α,α,α-Trifluoro-p-xylene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Methyl-4-(trifluoromethyl)benzene4-Methylbenzotrifluoride4-(Trifluoromethyl)toulene

L11043-03 L11043-06 L11043-03 L11043-06
AAAL11043-06EA 136.99 USD
CAAAAL11043-03 AAAL11043-06 CAAAAL11043-06 AAAL11043-03
α,α,α-Trifluoro-p-xylene 98%
α,α,α-Trifluoro-p-xylene
Formula: C₈H₇F₃
MW: 160.14 g/mol
Boiling Pt: 129 °C
Density: 1.144
Flash Pt: 27 °C(80 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075476
CAS Number: 6140-17-6
EINECS: 228-124-2
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR