6-Fluorosalicylic acid 96%

Danger

Synonyms: 2-Fluoro-6-hydroxybenzoic acid6-Fluoro-2-hydroxybenzoic acid

L10907-03 L10907-03
AAAL10907-03EA 120.23 USD
AAAL10907-03 CAAAAL10907-03
6-Fluorosalicylic acid 96%
6-Fluorosalicylic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
Melting Pt: 159…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00052179
CAS Number: 67531-86-6
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR