2,6-Diisopropylphenyl isothiocyanate 97%, Technical Grade

Danger

Synonyms: 1,3-Diisopropyl-2-isothiocyanatobenzene

L10893-03 L10893-06 L10893-06 L10893-03
AAAL10893-06EA 109.22 USD
CAAAAL10893-03 AAAL10893-06 AAAL10893-03 CAAAAL10893-06
2,6-Diisopropylphenyl isothiocyanate 97%, Technical Grade
2,6-Diisopropylphenyl isothiocyanate
Formula: C₁₃H₁₇NS
MW: 219.35 g/mol
Boiling Pt: 139…140 °C/5 mm
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041341
CAS Number: 25343-70-8
EINECS: 000-000-0
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR