2-Chloro-5-nitrotoluene 98+%

Warning

Synonyms: 1-Chloro-2-methyl-4-nitrobenzene

L10832-14 L10832-06 L10832-14 L10832-06
AAAL10832-06EA 39.47 USD
CAAAAL10832-06 AAAL10832-06 CAAAAL10832-14 AAAL10832-14
2-Chloro-5-nitrotoluene 98+%
2-Chloro-5-nitrotoluene
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 94…96 °C/4 mm
Melting Pt: 42…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00041430
CAS Number: 13290-74-9
EINECS: 236-306-8
UN: 3457
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR