Dinonyl sulfide 98%

L10638-14 L10638-14 L10638-06 L10638-06
AAAL10638-06EA 32.95 USD
AAAL10638-06 CAAAAL10638-06 AAAL10638-14 CAAAAL10638-14
Dinonyl sulfide 98%
Dinonyl sulphide
Formula: C₁₈H₃₈S
MW: 286.57 g/mol
Boiling Pt: 165…167 °C/3 mm
Melting Pt: 15 °C
Density: 0.855
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027342
CAS Number: 929-98-6
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR