2,3-Dichlorothiophene 97%

Danger

L10578-06 L10578-03 L10578-06 L10578-03
AAAL10578-03EA 52.99 USD
CAAAAL10578-03 AAAL10578-03 AAAL10578-06 CAAAAL10578-06
2,3-Dichlorothiophene 97%
2,3-Dichlorothiophene
Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –26.2 °C
Density: 1.470
Flash Pt: 70 °C(158 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043890
CAS Number: 17249-79-5
EINECS: 000-000-0
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR