2-Nitro-4-(trifluoromethyl)benzonitrile 98%

Warning

Synonyms: 4-Cyano-3-nitrobenzotrifluoride

L10557-14 L10557-06 L10557-06 L10557-14
AAAL10557-06EA 84.08 USD
CAAAAL10557-06 AAAL10557-06 CAAAAL10557-14 AAAL10557-14
2-Nitro-4-(trifluoromethyl)benzonitrile 98%
2-Nitro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₃F₃N₂O₂
Boiling Pt: 156…158 °C/18 mm
Melting Pt: 40…44 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014684
CAS Number: 778-94-9
EINECS: 212-298-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR