(R)-(-)-1-Phenyl-1,2-ethanediol 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
(R)-1-Phenylethane-1,2-diol
, (R)-(-)-Phenylethane-1,2-diol
, (R)-(-)-Phenyl-1,2-ethanediol
, (R)-(-)-α,β-Dihydroxyethylbenzene
L10494-MD
L10494-03
L10494-MD
L10494-03
AAAL10494-MDEA
30.04
USD
AAAL10494-MD
CAAAAL10494-MD
AAAL10494-03
CAAAAL10494-03
(R)-(-)-1-Phenyl-1,2-ethanediol 99%
(R)-(-)-1-Phenyl-1,2-ethanediol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 272…274 °C Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064262 CAS Number: 16355-00-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...