Dipropyl disulfide 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-DipropyldisulphanePropyl disulphide1,2-DipropyldisulfanePropyl disulfide

L09881-22 L09881-22
AAAL09881-22EA 77.79 USD
CAAAAL09881-22 AAAL09881-22
Dipropyl disulfide 99%
Dipropyl disulphide
Formula: C₆H₁₄S₂
MW: 150.31 g/mol
Boiling Pt: 193…194 °C
Melting Pt: –86 °C
Density: 0.960
Flash Pt: 64 °C(147 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009378
CAS Number: 629-19-6
EINECS: 211-079-8
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR