2-Amino-4,5-dimethoxybenzonitrile 98%

Warning

Synonyms: 2-Cyano-4,5-dimethoxyaniline

L09545-06 L09545-14 L09545-14 L09545-06
AAAL09545-06EA 66.64 USD
AAAL09545-06 CAAAAL09545-14 CAAAAL09545-06 AAAL09545-14
2-Amino-4,5-dimethoxybenzonitrile 98%
2-Amino-4,5-dimethoxybenzonitrile
Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
Melting Pt: 97…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00058682
CAS Number: 26961-27-3
EINECS: 000-000-0
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR