1-(3-Fluorophenyl)-2-thiourea 99%

Danger

Synonyms: N-(3-Fluorophenyl)thiourea3-Fluorophenylthiourea

L09538-03 L09538-06 L09538-03 L09538-06
AAAL09538-03EA 38.08 USD
CAAAAL09538-03 AAAL09538-03 CAAAAL09538-06 AAAL09538-06
1-(3-Fluorophenyl)-2-thiourea 99%
1-(3-Fluorophenyl)-2-thiourea
Formula: (C₆H₄F)NC(S)NH₂
MW: 170.21 g/mol
Melting Pt: 114…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00041167
CAS Number: 458-05-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR