1,1,2-Trichloro-2,3,3-trifluorocyclobutane 98%

Warning

L09469-06 L09469-06
AAAL09469-06EA 55.35 USD
CAAAAL09469-06 AAAL09469-06
1,1,2-Trichloro-2,3,3-trifluorocyclobutane 98%
1,1,2-Trichloro-2,3,3-trifluorocyclobutane
Formula: C₄H₂Cl₃F₃
MW: 213.41 g/mol
Boiling Pt: 119…120 °C
Melting Pt: –32 °C
Density: 1,594
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00039439
CAS Number: 697-17-6
EINECS: 211-804-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR