1-(4-Nitrophenyl)-2-thiourea 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Nitrophenyl)thioureaN-(4-Nitrophenyl)thiourea4-Nitrophenylthiourea

L09331-06 L09331-03 L09331-03 L09331-06
AAAL09331-03EA 29.19 USD
AAAL09331-03 CAAAAL09331-03 AAAL09331-06 CAAAAL09331-06
1-(4-Nitrophenyl)-2-thiourea 98%
1-(4-Nitrophenyl)-2-thiourea
Formula: (C₆H₄(NO₂))NC(S)NH₂
MW: 197.22 g/mol
Melting Pt: ca 207 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007304
CAS Number: 3696-22-8
EINECS: 223-021-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR