2,3-Dichlorooctafluorobutane 97%

L09252-06 L09252-06 L09252-03 L09252-03
AAAL09252-03EA 26.76 USD
CAAAAL09252-03 CAAAAL09252-06 AAAL09252-03 AAAL09252-06
2,3-Dichlorooctafluorobutane 97%
2,3-Dichlorooctafluorobutane
Formula: C₄Cl₂F₈
MW: 270.94 g/mol
Boiling Pt: 62…63 °C
Melting Pt: ca …68 °C
Density: 1.680
Storage Temperature: Ambient
MDL Number: MFCD00018064
CAS Number: 355-20-4
EINECS: 206-578-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR