4-Nitroindole 97%

Warning

Synonyms: 4-Nitro-1H-indole

L09177-MC L09177-MC L09177-03 L09177-ME L09177-03 L09177-ME
AAAL09177-MEEA 92.4 USD
AAAL09177-ME CAAAAL09177-MC AAAL09177-MC CAAAAL09177-ME CAAAAL09177-03 AAAL09177-03
4-Nitroindole 97%
4-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 202…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00010056
CAS Number: 4769-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR