Butylcyclohexane 99%

Supplier: Thermo Scientific Chemicals
Warning

L08845-18 L08845-18 L08845-09 L08845-09
AAAL08845-18EA 102.99 USD
CAAAAL08845-18 AAAL08845-18 CAAAAL08845-09 AAAL08845-09
Butylcyclohexane 99%
Butylcyclohexane
Formula: C₁₀H₂₀
Boiling Pt: 178…180 °C
Melting Pt: –78 °C
Density: 0.818
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001530
CAS Number: 1678-93-9
EINECS: 216-837-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR