3,4-Diethoxybenzoic acid 99%

Warning

L08717-14 L08717-06 L08717-14 L08717-06
AAAL08717-06EA 18.12 USD
AAAL08717-06 CAAAAL08717-06 AAAL08717-14 CAAAAL08717-14
3,4-Diethoxybenzoic acid 99%
3,4-Diethoxybenzoic acid
Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00002504
CAS Number: 5409-31-4
EINECS: 226-478-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR