2-Fluoro-5-iodotoluene 99%

Warning

Synonyms: 2-Fluoro-5-iodo-1-methylbenzene1-Fluoro-4-iodo-2-methylbenzene

L08396-03 L08396-06 L08396-03 L08396-06
AAAL08396-06EA 196.47 USD
CAAAAL08396-06 AAAL08396-06 CAAAAL08396-03 AAAL08396-03
2-Fluoro-5-iodotoluene 99%
2-Fluoro-5-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
Boiling Pt: 86…87 °C/9 mm
Storage Temperature: Ambient
MDL Number: MFCD00013710
CAS Number: 452-68-6
EINECS: 207-206-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR