2-Hydroxyethylmesitylene 98%

Synonyms: 1-(2-Hydroxyethyl)-2,4,6-trimethylbenzene2-(2,4,6-Trimethylphenyl)ethanol2-Hydroxyethylmesitylene2,4,6-Trimethylphenethyl alcohol2-Mesitylethanol

L08032-06 L08032-06 L08032-03 L08032-14 L08032-03 L08032-14
AAAL08032-06EA 102.96 USD
AAAL08032-06 CAAAAL08032-06 AAAL08032-03 AAAL08032-14 CAAAAL08032-14 CAAAAL08032-03
2-Hydroxyethylmesitylene 98%
2-Hydroxyethylmesitylene
Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00020609
CAS Number: 6950-92-1
EINECS: 230-123-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR