Nitrocyclopentane 99%

Supplier: Thermo Scientific Chemicals
Warning

L08029-03 L08029-14 L08029-06 L08029-06 L08029-03 L08029-14
AAAL08029-03EA 56.79 USD
AAAL08029-03 AAAL08029-14 CAAAAL08029-06 AAAL08029-06 CAAAAL08029-03 CAAAAL08029-14
Nitrocyclopentane 99%
Nitrocyclopentane
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 179…180 °C
Density: 1.082
Flash Pt: 67 °C(152 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001362
CAS Number: 2562-38-1
EINECS: 219-884-6
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR