4-Ethylbenzonitrile 98%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Ethylbenzonitrile

L07921-09 L07921-18 L07921-18 L07921-09
AAAL07921-18EA 421.3 USD
CAAAAL07921-09 CAAAAL07921-18 AAAL07921-18 AAAL07921-09
4-Ethylbenzonitrile 98%
4-Ethylbenzonitrile
Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 237 °C
Density: 0,956
Flash Pt: 97 °C(206 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016383
CAS Number: 25309-65-3
EINECS: 246-811-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR