1-Nitro-4-propylbenzene 96%

Supplier: Thermo Scientific Chemicals
Warning

L07730-03 L07730-14
AAL07730-03EA 34.6 USD
AAL07730-03 AAL07730-14
1-Nitro-4-propylbenzene 96%
1-Nitro-4-propylbenzene
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 96…100 °C/2 mm
Melting Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00051839
CAS Number: 10342-59-3
EINECS: 233-743-6
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR