2-Chloro-6-fluorocinnamic acid predominantly trans 98%

Warning

Synonyms: (2E)-3-(2-Chloro-6-fluorophenyl)acrylic acid3-(2-Chloro-6-fluorophenyl)acrylic acid

L07468-03 L07468-06 L07468-03 L07468-06
AAAL07468-03EA 21.35 USD
CAAAAL07468-03 CAAAAL07468-06 AAAL07468-03 AAAL07468-06
2-Chloro-6-fluorocinnamic acid predominantly trans 98%
2-Chloro-6-fluorocinnamic acid
Formula: C₉H₆ClFO₂
MW: 200.6 g/mol
Melting Pt: 164…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00051582
CAS Number: 392-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR