3,5-Dibromoanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-3,5-dibromobenzoicacid2-Amino-3,5-dibromobenzoic acid

L07419-06 L07419-06 L07419-14 L07419-14
AAAL07419-06EA 36.19 USD
AAAL07419-06 CAAAAL07419-06 AAAL07419-14 CAAAAL07419-14
3,5-Dibromoanthranilic acid 98%
3,5-Dibromoanthranilic acid
Formula: C₇H₅Br₂NO₂
MW: 294.93 g/mol
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00017087
CAS Number: 609-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR