4-Fluoro-2-iodotoluene 98%

Warning

Synonyms: 1-Fluoro-3-iodo-4-methylbenzene4-Fluoro-2-iodo-1-methylbenzene

L06833-06 L06833-14 L06833-06 L06833-14
AAAL06833-14EA 268.52 USD
CAAAAL06833-14 CAAAAL06833-06 AAAL06833-14 AAAL06833-06
4-Fluoro-2-iodotoluene 98%
4-Fluoro-2-iodotoluene
Formula: C₇H₆FI
MW: 236.03 g/mol
Boiling Pt: 206…208 °C
Density: 1.81
Flash Pt: 87 °C(189 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013709
CAS Number: 13194-67-7
EINECS: 236-153-7
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR