9-Bromoanthracene 96%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

L06733-06 L06733-06 L06733-03 L06733-03
AAAL06733-06EA 68.76 USD
AAAL06733-06 CAAAAL06733-03 CAAAAL06733-06 AAAL06733-03
9-Bromoanthracene 96%, Technical Grade
9-Bromoanthracene
Formula: C₁₄H₉Br
Melting Pt: 97…103 °C
Storage Temperature: Ambient
MDL Number: MFCD00001243
CAS Number: 1564-64-3
EINECS: 216-359-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR