8-Quinolinesulfonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
8-Quinolinesulphonic acid
, 8-Quinolinesulfonic acid
L06640-18
L06640-18
AAAL06640-18EA
269.82
USD
CAAAAL06640-18
AAAL06640-18
8-Quinolinesulfonic acid 98%
8-Quinolinesulphonic acid
Formula:
C₉H₇NO₃S MW: 209.23 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00024037 CAS Number: 85-48-3 EINECS: 201-611-7 UN: 2585 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...