Sodium nonyl sulfate 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nonyl sulphuric acid saltNonyl sulfuric acid salt

L06418-06 L06418-03 L06418-06 L06418-03
AAAL06418-03EA 43.69 USD
CAAAAL06418-03 CAAAAL06418-06 AAAL06418-03 AAAL06418-06
Sodium nonyl sulfate 99%
Sodium nonyl sulphate
Formula: C₉H₁₉NaO₄S
MW: 246.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024893
CAS Number: 1072-15-7
EINECS: 214-002-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR