Sodium hexadecyl sulfate 99%

Warning

Synonyms: Hexadecyl sulfuric acid saltHexadecyl sulphuric acid salt

L06402-06 L06402-03 L06402-03
AAAL06402-03EA 69.85 USD
CAAAAL06402-06 AAAL06402-03 CAAAAL06402-03
Sodium hexadecyl sulfate 99%
Sodium hexadecyl sulphate
Formula: C₁₆H₃₃NaO₄S
MW: 344.49 g/mol
Melting Pt: 187…192 °C
MDL Number: MFCD00047766
CAS Number: 1120-01-0
EINECS: 214-292-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR