4-Chlorobutyrophenone 95%

Synonyms: ω-Chlorobutyrophenoneγ-Chlorobutyrophenone

L06383-14 L06383-06 L06383-06 L06383-14
AAAL06383-14EA 158.66 USD
CAAAAL06383-14 AAAL06383-14 CAAAAL06383-06 AAAL06383-06
4-Chlorobutyrophenone 95%
4-Chlorobutyrophenone
Formula: (C₆H₅)C(O)(CH₂)₃Cl
MW: 182.65 g/mol
Boiling Pt: 138…140 °C/5 mm
Melting Pt: 19…20 °C
Density: 1.137
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001005
CAS Number: 939-52-6
EINECS: 213-362-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR