4-Nitro-3-picoline-N-oxide 98%

Warning

Synonyms: 3-Methyl-4-nitropyridine N-oxide4-Nitro-3-picoline N-oxide4-Nitropicoline N-oxide3-Methyl-4-nitropyridine 1-oxide3-Methyl-4-nitropyridine-1-oxide3-Methyl-4-nitropyridine-N-oxide

L06099-03 L06099-06 L06099-06 L06099-03
AAAL06099-03EA 60.88 USD
CAAAAL06099-06 AAAL06099-03 AAAL06099-06 CAAAAL06099-03
4-Nitro-3-picoline-N-oxide 98%
4-Nitro-3-picoline N-oxide
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00014626
CAS Number: 1074-98-2
EINECS: 214-050-8
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR