α,α,α-Trifluoro-3-nitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Nitrobenzotrifluoride

L05948-22 L05948-22
AAAL05948-22EA 100.99 USD
AAAL05948-22 CAAAAL05948-22
α,α,α-Trifluoro-3-nitrotoluene 97%
α,α,α-Trifluoro-3-Nitrotoluene
Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Boiling Pt: 201…205 °C
Melting Pt: –5 °C
Density: 1.435
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007260
CAS Number: 98-46-4
EINECS: 202-670-1
UN: 2306
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR