1-(2,5-Xylyl)piperazin 96%

Danger

Synonyms: 1-(2,5-Dimethylphenyl)piperazine

L05744-06 L05744-06 L05744-14 L05744-14
AAAL05744-06EA 58.21 USD
CAAAAL05744-14 AAAL05744-06 AAAL05744-14 CAAAAL05744-06
1-(2,5-Xylyl)piperazin 96%
1-(2,5-Xylyl)piperazin
Formula: C₁₂H₁₈N₂
MW: 190.29 g/mol
Boiling Pt: 154 °C/10 mm
Melting Pt: 42…47 °C
Storage Temperature: Ambient
MDL Number: MFCD00038378
CAS Number: 1013-25-8
EINECS: 213-795-6
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR