3-Benzyloxy-p-anisic acid 97+%

Warning

Synonyms: 3-(Benzyloxy)-4-methoxybenzoic acid3-Benzyloxy-4-methoxybenzoic acid4-Methoxy-3-benzyloxybenzoic acid

L05731-03 L05731-06 L05731-14 L05731-03 L05731-06 L05731-14
AAAL05731-06EA 151.11 USD
CAAAAL05731-06 AAAL05731-06 AAAL05731-03 AAAL05731-14 CAAAAL05731-03 CAAAAL05731-14
3-Benzyloxy-p-anisic acid 97+%
3-Benzyloxy-p-anisic acid
Formula: C₁₅H₁₄O₄
MW: 258.27 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00016526
CAS Number: 58452-00-9
EINECS: 261-260-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR