3-Ethoxybenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L05721-09
L05721-09
AAAL05721-09EA
105.99
USD
CAAAAL05721-09
AAAL05721-09
3-Ethoxybenzoic acid 98+%
3-Ethoxybenzoic acid
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Melting Pt: 135…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016517 CAS Number: 621-51-2 EINECS: 210-690-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...