2,6-Dichlorocinnamic acid predominantly trans 98%

Warning

Synonyms: (2E)-3-(2,6-Dichlorophenyl)acrylic acid3-(2,6-Dichlorophenyl)acrylic acid

L05614-14 L05614-06 L05614-06 L05614-14
AAAL05614-14EA 114.33 USD
AAAL05614-14 CAAAAL05614-14 AAAL05614-06 CAAAAL05614-06
2,6-Dichlorocinnamic acid predominantly trans 98%
2,6-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
Boiling Pt: 333 °C (1013 hPa)
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00004374
CAS Number: 5345-89-1
EINECS: 226-301-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR