n-Triacontane 99%
Supplier: Thermo Scientific Chemicals
L05386-03
L05386-06
L05386-03
L05386-MD
L05386-06
L05386-MD
AAAL05386-03EA
62.19
USD
AAAL05386-03
CAAAAL05386-03
AAAL05386-MD
AAAL05386-06
CAAAAL05386-MD
CAAAAL05386-06
n-Triacontane 99%
n-Triacontane
Formula:
H₃C(CH₂)₂₈CH₃ MW: 422.82 g/mol Boiling Pt: 455 °C Melting Pt: 65…68 °C Density: 0.7797 g/cm³ (20 °C) Flash Pt: 237 °C(458 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009410 CAS Number: 638-68-6 EINECS: 211-349-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...