4-Methyl-1-pentyne-3-ol 97%

Warning

Synonyms: 3-Hydroxy-4-methyl-1-pentyne4-Methylpent-1-yn-3-ol

L05318-18 L05318-18 L05318-09 L05318-09
AAAL05318-18EA 449.17 USD
CAAAAL05318-18 AAAL05318-18 CAAAAL05318-09 AAAL05318-09
4-Methyl-1-pentyne-3-ol 97%
4-Methyl-1-pentyne-3-ol
Formula: C₆H₁₀O
MW: 98.14 g/mol
Boiling Pt: 133…134 °C
Density: 0.929
Flash Pt: 41 °C(105 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039847
CAS Number: 565-68-4
EINECS: 209-287-9
UN: 1987
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR