Hexaphenyldisilane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,1,2,2,2-Hexaphenyldisilane

L04928-03 L04928-06 L04928-06 L04928-03
AAAL04928-06EA 157.99 USD
CAAAAL04928-03 AAAL04928-06 AAAL04928-03 CAAAAL04928-06
Hexaphenyldisilane 98%
Hexaphenyldisilane
Formula: C₃₆H₃₀Si₂
MW: 518.81 g/mol
Melting Pt: 368…370 °C
Storage Temperature: Ambient
MDL Number: MFCD00039558
CAS Number: 1450-23-3
EINECS: 215-913-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR