Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

2-(2-Chlorophenoxy)ethanol 98%

Supplier: Alfa Aesar
Warning

L04368-14 L04368-22 L04368-14 L04368-06 L04368-22 L04368-06
AAAL04368-14EA 93.66 USD InStock
AAAL04368-14 AAAL04368-22 CAAAAL04368-14 CAAAAL04368-06 CAAAAL04368-22 AAAL04368-06
2-(2-Chlorophenoxy)ethanol 98%
2-(2-Chlorophenoxy)ethanol


Formula: C₈H₉ClO₂
MW: 172.61 g/mol
Boiling Pt: 114…116 °C (1 mmHg)
Density: 1.2567 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014034
CAS Number: 15480-00-9
EINECS: 239-501-6
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty