Methyl butyrate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Butyric acid methyl esterButanoic acid methyl ester

L04250-AP L04250-AP
AAAL04250-APEA 60.39 USD
AAAL04250-AP CAAAAL04250-AP
Methyl butyrate 98+%
Methyl butyrate
Formula: C₅H₁₀O₂
Boiling Pt: 100…102 °C
Melting Pt: –85 to –84 °C
Density: 0.898
Flash Pt: 11 °C(51 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009391
CAS Number: 623-42-7
EINECS: 210-792-1
UN: 1237
ADR: "3",II
Merck Index: 13,06062

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR