3-Fluorobenzal chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: α,α-Dichloro-3-fluortoluene

L04121-14 L04121-14 L04121-06 L04121-06
AAAL04121-06EA 68.69 USD
CAAAAL04121-06 AAAL04121-06 CAAAAL04121-14 AAAL04121-14
3-Fluorobenzal chloride 98%
3-Fluorobenzal chloride
Formula: C₇H₅Cl₂F
MW: 179.02 g/mol
Boiling Pt: 195…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00037112
CAS Number: 402-64-2
EINECS: 206-952-5
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR