2,3,4-Tribromothiophene 97%
|
Warning
|
L03946-03
L03946-03
L03946-06
L03946-06
AAAL03946-03EA
65.44
USD
AAAL03946-03
CAAAAL03946-03
AAAL03946-06
CAAAAL03946-06
2,3,4-Tribromothiophene 97%
2,3,4-Tribromothiophene
Formula:
C₄HBr₃S MW: 320.83 g/mol Boiling Pt: 150…152 °C/20 mm Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041210 CAS Number: 3141-25-1 EINECS: 221-545-2 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...